N'-(4-Fluorobenzyl)ethane-1,2-diamine, N-BOC protected
Supplier: Apollo Scientific
Synonyms: tert-Butyl {2-[(4-fluorobenzyl)amino]ethyl}carbamate
PC400513-1G PC400513-500MG
APOSPC400513-1GEA 649 EUR
APOSPC400513-1G APOSPC400513-500MG
N'-(4-Fluorobenzyl)ethane-1,2-diamine, N-BOC protected
N'-(4-Fluorobenzyl)ethane-1,2-diamine, N-BOC protected
Formula: C₁₄H₂₁FN₂O₂
MW: 268.327 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22690784
CAS Number: 519172-79-3