3-Methyl-1-[2-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine
Supplier: Apollo Scientific
Warning
Synonyms: 3-Methyl-1-(2-(trifluoromethyl)phenyl)-1H-pyrazol-5-amine, 5-Amino-3-methyl-1-[2-(trifluoromethyl)phenyl]-1H-pyrazole, 2-(5-Amino-3-methyl-1H-pyrazol-1-yl)benzotrifluoride
PC400534-1G PC400534-250MG
APOSPC400534-1GEA 163 EUR
APOSPC400534-1G APOSPC400534-250MG
3-Methyl-1-[2-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine
3-Methyl-1-[2-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine
Formula: C₁₁H₁₀F₃N₃
MW: 241.212 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02715834
CAS Number: 380238-10-8