8-Chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine-3-thiol
Supplier: Apollo Scientific
PC401036-1G PC401036-5G
APOSPC401036-1GEA 277 EUR
APOSPC401036-1G APOSPC401036-5G
8-Chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine-3-thiol
8-Chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine-3-thiol
Formula: C₇H₃ClF₃N₃S
MW: 253.632 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04610459
CAS Number: 571917-27-6