2-Cyano-3-phenoxy-5-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Warning
PC401044-1G PC401044-250MG
APOSPC401044-1GEA 241 EUR
APOSPC401044-1G APOSPC401044-250MG
2-Cyano-3-phenoxy-5-(trifluoromethyl)pyridine
2-Cyano-3-phenoxy-5-(trifluoromethyl)pyridine
Formula: C₁₃H₇F₃N₂O
MW: 264.203 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25970580
CAS Number: 1065607-70-6