(S)-3-(4-Fluorophenyl)piperidine hydrochloride
Supplier: Apollo Scientific
PC402143-1G PC402143-250MG
APOSPC402143-1GEA 877 EUR
APOSPC402143-1G APOSPC402143-250MG
(S)-3-(4-Fluorophenyl)piperidine hydrochloride
(S)-3-(4-Fluorophenyl)piperidine hydrochloride
Formula: C₁₁H₁₄FN·ClH
MW: 215.695 g/mol
Storage Temperature: Refrigerator
CAS Number: 1217829-49-6