5-Iodo-3-(trifluoromethyl)-1H-pyrazole
Supplier: Apollo Scientific
Warning
Synonyms: 3-Iodo-5-(trifluoromethyl)-1H-pyrazole
PC404013-1G PC404013-250MG PC404013-5G
APOSPC404013-1GEA 223 EUR
APOSPC404013-1G APOSPC404013-250MG APOSPC404013-5G
5-Iodo-3-(trifluoromethyl)-1H-pyrazole
5-Iodo-3-(trifluoromethyl)-1H-pyrazole
Formula: C₄H₂F₃IN₂
MW: 261.972 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24540492
CAS Number: 1015780-53-6