trans-tert-Butyl 3-(4-trifluoromethylphenyl)-aziridine-2-carboxylate
Supplier: Apollo Scientific
PC404510-1G PC404510-250MG
APOSPC404510-1GEA 525 EUR
APOSPC404510-1G APOSPC404510-250MG
trans-tert-Butyl 3-(4-trifluoromethylphenyl)-aziridine-2-carboxylate
trans-tert-Butyl 3-(4-trifluoromethylphenyl)-aziridine-2-carboxylate
Formula: C₁₄H₁₆F₃NO₂
MW: 287.278 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26936312
CAS Number: 1980008-32-9