trans-tert-Butyl 3-(3-fluorophenyl)-aziridine-2-carboxylate
Supplier: Apollo Scientific
PC404512-1G PC404512-250MG
APOSPC404512-1GEA 509 EUR
APOSPC404512-1G APOSPC404512-250MG
trans-tert-Butyl 3-(3-fluorophenyl)-aziridine-2-carboxylate
trans-tert-Butyl 3-(3-fluorophenyl)-aziridine-2-carboxylate
Formula: C₁₃H₁₆FNO₂
Storage Temperature: Ambient
MDL Number: MFCD26936314
CAS Number: 1980007-30-4