1-[4-(Trifluoromethoxy)phenyl]cyclopropanecarboxylic acid
Supplier: Apollo Scientific
Warning
PC404538-1G PC404538-250MG
APOSPC404538-1GEA 248 EUR
APOSPC404538-1G APOSPC404538-250MG
1-[4-(Trifluoromethoxy)phenyl]cyclopropanecarboxylic acid
1-[4-(Trifluoromethoxy)phenyl]cyclopropanecarboxylic acid
Formula: C₁₁H₉F₃O₃
MW: 246.183 g/mol
Melting Pt: 104…105 °C
Storage Temperature: Ambient
MDL Number: MFCD11037007
CAS Number: 936727-93-4