(2,8-Bis-trifluoromethyl-quinolin-4-yl)-pyridin-2-yl-methanone
Supplier: Apollo Scientific
PC404544-1G PC404544-5G
APOSPC404544-1GEA 301 EUR
APOSPC404544-1G APOSPC404544-5G
(2,8-Bis-trifluoromethyl-quinolin-4-yl)-pyridin-2-yl-methanone
(2,8-Bis-trifluoromethyl-quinolin-4-yl)-pyridin-2-yl-methanone
Formula: C₁₇H₈F₆N₂O
MW: 370.249 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05863551
CAS Number: 35853-55-5