2-Chloro-7-fluoroquinoline-3-carboxaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-7-fluoroquinoline-3-carbaldehyde
PC404559-250MG PC404559-1G
APOSPC404559-250MGEA 145 EUR
APOSPC404559-250MG APOSPC404559-1G
2-Chloro-7-fluoroquinoline-3-carboxaldehyde
2-Chloro-7-fluoroquinoline-3-carboxaldehyde
Formula: C₁₀H₅ClFNO
MW: 209.604 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09787681
CAS Number: 745830-16-4