2,8-Bis(trifluoromethyl)-4-(piperazin-1-yl)quinoline
Supplier: Apollo Scientific
Warning
PC407010-100MG PC407010-1G PC407010-250MG
APOSPC407010-100MGEA 79.9 EUR
APOSPC407010-100MG APOSPC407010-1G APOSPC407010-250MG
2,8-Bis(trifluoromethyl)-4-(piperazin-1-yl)quinoline
2,8-Bis(trifluoromethyl)-4-(piperazin-1-yl)quinoline
Formula: C₁₅H₁₃F₆N₃
MW: 349.274 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07700226
CAS Number: 401567-76-8