4-Chloro-8-trifluoromethoxyquinoline
Supplier: Apollo Scientific
Synonyms: 4-Chloro-8-(trifluoromethoxy)quinoline
PC407052-1G PC407052-250MG
APOSPC407052-1GEA 260 EUR
APOSPC407052-1G APOSPC407052-250MG
4-Chloro-8-trifluoromethoxyquinoline
4-Chloro-8-trifluoromethoxyquinoline
Formula: C₁₀H₅ClF₃NO
MW: 247.601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861865
CAS Number: 40516-42-5