Perfluoro(2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecanoic) acid
Supplier: Apollo Scientific
Danger
PC408088-1G PC408088-5G
APOSPC408088-1GEA 109 EUR
APOSPC408088-1G APOSPC408088-5G
Perfluoro(2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecanoic) acid
Perfluoro(2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecanoic) acid
Formula: C₁₅HF₂₉O₆
MW: 828.119 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03412274
CAS Number: 65150-95-0
UN: 3265
ADR: 8,III