2-Fluoro-3-methoxybenzylamine
Supplier: Apollo Scientific
Danger
Synonyms: (2-Fluoro-3-methoxyphenyl)methylamine
PC408258-1G PC408258-5G
APOSPC408258-1GEA 108 EUR
APOSPC408258-1G APOSPC408258-5G
2-Fluoro-3-methoxybenzylamine
2-Fluoro-3-methoxybenzylamine
Formula: C₈H₁₀FNO
MW: 155.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11045854
CAS Number: 93071-81-9
UN: 3267
ADR: 8,III