5-Fluoro-2-methylpyrimidin-4-amine
Supplier: Apollo Scientific
PC408553-1G PC408553-250MG
APOSPC408553-1GEA 328 EUR
APOSPC408553-1G APOSPC408553-250MG
5-Fluoro-2-methylpyrimidin-4-amine
5-Fluoro-2-methylpyrimidin-4-amine
Formula: C₅H₆FN₃
MW: 127.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23099431
CAS Number: 1416372-67-2