4-(Difluoromethoxy)-2-methylaniline hydrochloride
Supplier: Apollo Scientific
Warning
PC410009-1G PC410009-5G
APOSPC410009-1GEA 66.7 EUR
APOSPC410009-1G APOSPC410009-5G
4-(Difluoromethoxy)-2-methylaniline hydrochloride
4-(Difluoromethoxy)-2-methylaniline hydrochloride
Formula: C₈H₉F₂NO·ClH
MW: 209.621 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04967256
CAS Number: 1185175-00-1