1-(2,2-Difluoroethyl)piperazine 95%
Supplier: Apollo Scientific
Warning
PC410047-1G PC410047-250MG PC410047-5G
APOSPC410047-1GEA 173 EUR
APOSPC410047-1G APOSPC410047-250MG APOSPC410047-5G
1-(2,2-Difluoroethyl)piperazine 95%
1-(2,2-Difluoroethyl)piperazine
Formula: C₆H₁₂F₂N₂
MW: 150.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08696507
CAS Number: 767609-14-3