4-(2,2-Difluoroethoxy)aniline hydrochloride
Supplier: Apollo Scientific
Warning
PC410078-1G PC410078-5G
APOSPC410078-1GEA 60.9 EUR
APOSPC410078-1G APOSPC410078-5G
4-(2,2-Difluoroethoxy)aniline hydrochloride
4-(2,2-Difluoroethoxy)aniline hydrochloride
Formula: C₈H₉F₂NO·ClH
MW: 209.621 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16621779
CAS Number: 1431965-53-5