2-(Difluoromethoxy)-4-methylaniline hydrochloride
Supplier: Apollo Scientific
Warning
PC410124-1G PC410124-5G
APOSPC410124-1GEA 87 EUR
APOSPC410124-1G APOSPC410124-5G
2-(Difluoromethoxy)-4-methylaniline hydrochloride
2-(Difluoromethoxy)-4-methylaniline hydrochloride
Formula: C₈H₉F₂NO·ClH
MW: 209.621 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25371273
CAS Number: 1431962-99-0