1-(3,3-Difluorocyclobutyl)ethan-1-ol
Supplier: Apollo Scientific
PC411282-1G PC411282-250MG PC411282-500MG
APOSPC411282-1GEA 877 EUR
APOSPC411282-1G APOSPC411282-250MG APOSPC411282-500MG
1-(3,3-Difluorocyclobutyl)ethan-1-ol
1-(3,3-Difluorocyclobutyl)ethan-1-ol
Formula: C₆H₁₀F₂O
MW: 136.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29918535
CAS Number: 1784303-38-3