1-(Difluoromethyl)-1H-pyrazole
Supplier: Apollo Scientific
Warning
PC411300-1G PC411300-5G
APOSPC411300-1GEA 122 EUR
APOSPC411300-1G APOSPC411300-5G
1-(Difluoromethyl)-1H-pyrazole
1-(Difluoromethyl)-1H-pyrazole
Formula: C₄H₄F₂N₂
MW: 118.085 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04971168
CAS Number: 908014-77-7
UN: 1993
ADR: 3 III