1-(Difluoromethyl)-1H-pyrazol-4-amine hydrochloride
Supplier: Apollo Scientific
Warning
PC411304-1G PC411304-5G
APOSPC411304-1GEA 30.6 EUR
APOSPC411304-1G APOSPC411304-5G
1-(Difluoromethyl)-1H-pyrazol-4-amine hydrochloride
1-(Difluoromethyl)-1H-pyrazol-4-amine hydrochloride
Formula: C₄H₅F₂N₃·ClH
MW: 169.56 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14705743
CAS Number: 1221726-31-3