1-(2-Fluoroethyl)-4-iodo-5-methyl-1H-pyrazole
Supplier: Apollo Scientific
Warning
PC411306-1G PC411306-5G
APOSPC411306-1GEA 34.8 EUR
APOSPC411306-1G APOSPC411306-5G
1-(2-Fluoroethyl)-4-iodo-5-methyl-1H-pyrazole
1-(2-Fluoroethyl)-4-iodo-5-methyl-1H-pyrazole
Formula: C₆H₈FIN₂
MW: 254.044 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25949457
CAS Number: 1443278-91-8