1-(2,2-Difluoroethyl)-4-methyl-1H-pyrazol-3-amine
Supplier: Apollo Scientific
Warning
PC411332-1G PC411332-5G
APOSPC411332-1GEA 66.7 EUR
APOSPC411332-1G APOSPC411332-5G
1-(2,2-Difluoroethyl)-4-methyl-1H-pyrazol-3-amine
1-(2,2-Difluoroethyl)-4-methyl-1H-pyrazol-3-amine
Formula: C₆H₉F₂N₃
MW: 161.153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11215400
CAS Number: 1174875-53-6