3-(Difluoromethyl)-1H-pyrazole-5-carboxylic acid
Supplier: Apollo Scientific
Warning
PC411335-1G PC411335-5G
APOSPC411335-1GEA 201 EUR
APOSPC411335-1G APOSPC411335-5G
3-(Difluoromethyl)-1H-pyrazole-5-carboxylic acid
3-(Difluoromethyl)-1H-pyrazole-5-carboxylic acid
Formula: C₅H₄F₂N₂O₂
MW: 162.094 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18262323
CAS Number: 681034-51-5