N-(4-Fluorophenyl)-3-(trifluoromethyl)benzamide 98%
Supplier: Apollo Scientific
Synonyms: 4'-Fluoro-3-(trifluoromethyl)benzanilide
PC4175W-1G PC4175W-5G
APOSPC4175W-1GEA 91.9 EUR
APOSPC4175W-1G APOSPC4175W-5G
N-(4-Fluorophenyl)-3-(trifluoromethyl)benzamide 98%
N-(4-Fluorophenyl)-3-(trifluoromethyl)benzamide
Formula: C₁₄H₉F₄NO
Melting Pt: 101…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00045089
CAS Number: 33489-71-3