4'-Chloro-2-(4-fluorophenyl)acetophenone
Supplier: Apollo Scientific
Danger
Synonyms: 1-(4-Chlorophenyl)-2-(4-fluorophenyl)ethanone
PC421014-1G PC421014-250MG PC421014-5G
APOSPC421014-1GEA 139 EUR
APOSPC421014-1G APOSPC421014-250MG APOSPC421014-5G
4'-Chloro-2-(4-fluorophenyl)acetophenone
4'-Chloro-2-(4-fluorophenyl)acetophenone
Formula: C₁₄H₁₀ClFO
MW: 248.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02063016
CAS Number: 126866-15-7