2-Chloro-3-(trifluoromethoxy)pyridine
Supplier: Apollo Scientific
PC421032-1G PC421032-500MG
APOSPC421032-1GEA 343 EUR
APOSPC421032-1G APOSPC421032-500MG
2-Chloro-3-(trifluoromethoxy)pyridine
2-Chloro-3-(trifluoromethoxy)pyridine
Formula: C₆H₃ClF₃NO
MW: 197.542 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17168453
CAS Number: 1206980-39-3