2-Bromo-3-fluoro-5-methylaniline 97%
Supplier: Apollo Scientific
Warning
PC421121-1G PC421121-250MG
APOSPC421121-1GEA 473 EUR
APOSPC421121-1G APOSPC421121-250MG
2-Bromo-3-fluoro-5-methylaniline 97%
2-Bromo-3-fluoro-5-methylaniline
Formula: C₇H₇BrFN
MW: 204.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28739689
CAS Number: 1805421-04-8