(S)-2-Chloro-1-(3,4-difluorophenyl)ethanol 99%
Supplier: Apollo Scientific
Danger
PC421194-1G PC421194-250MG PC421194-5G
APOSPC421194-1GEA 29 EUR
APOSPC421194-1G APOSPC421194-250MG APOSPC421194-5G
(S)-2-Chloro-1-(3,4-difluorophenyl)ethanol 99%
(S)-2-Chloro-1-(3,4-difluorophenyl)ethanol
Formula: C₈H₇ClF₂O
MW: 192.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09863590
CAS Number: 1006376-60-8