1-(6-Bromo-3-fluoropyridin-2-yl)ethanone 97%
Supplier: Apollo Scientific
PC421197-100MG PC421197-1G PC421197-250MG PC421197-5G
APOSPC421197-100MGEA 234 EUR
APOSPC421197-100MG APOSPC421197-1G APOSPC421197-250MG APOSPC421197-5G
1-(6-Bromo-3-fluoropyridin-2-yl)ethanone 97%
1-(6-Bromo-3-fluoropyridin-2-yl)ethanone
Formula: C₇H₅BrFNO
Storage Temperature: Ambient
MDL Number: MFCD18258632
CAS Number: 1016228-01-5