Methyl 2-(3,5-difluorophenyl)-2-hydroxyacetate 95%
Supplier: Apollo Scientific
PC421231-100MG PC421231-1G PC421231-250MG
APOSPC421231-100MGEA 252 EUR
APOSPC421231-100MG APOSPC421231-1G APOSPC421231-250MG
Methyl 2-(3,5-difluorophenyl)-2-hydroxyacetate 95%
Methyl 2-(3,5-difluorophenyl)-2-hydroxyacetate
Formula: C₉H₈F₂O₃
MW: 202.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11977773
CAS Number: 208259-37-4