2,2-Difluoropentanedioic acid
Supplier: Apollo Scientific
PC430166-1G PC430166-250MG
APOSPC430166-1GEA 765 EUR
APOSPC430166-1G APOSPC430166-250MG
2,2-Difluoropentanedioic acid
2,2-Difluoropentanedioic acid
Formula: C₅H₆F₂O₄
MW: 168.096 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20640982
CAS Number: 380-86-9