tert-Butyl 3,3-difluoro-4-hydroxypyrrolidine-1-carboxylate
Supplier: Apollo Scientific
PC430237-250MG PC430237-1G
APOSPC430237-250MGEA 285 EUR
APOSPC430237-250MG APOSPC430237-1G
tert-Butyl 3,3-difluoro-4-hydroxypyrrolidine-1-carboxylate
tert-Butyl 3,3-difluoro-4-hydroxypyrrolidine-1-carboxylate
Formula: C₉H₁₅F₂NO₃
MW: 223.217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27956892
CAS Number: 1434141-81-7