3,4-Difluoro-5-methylaniline
Supplier: Apollo Scientific
Warning
PC430267-250MG PC430267-100MG PC430267-1G
APOSPC430267-250MGEA 174 EUR
APOSPC430267-250MG APOSPC430267-100MG APOSPC430267-1G
3,4-Difluoro-5-methylaniline
3,4-Difluoro-5-methylaniline
Formula: C₇H₇F₂N
MW: 143.134 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22490569
CAS Number: 1505944-46-6