tert-Butyl 3,3-difluoro-4-hydroxypiperidine-1-carboxylate
Supplier: Apollo Scientific
Warning
PC430294-1G PC430294-250MG
APOSPC430294-1GEA 200 EUR
APOSPC430294-1G APOSPC430294-250MG
tert-Butyl 3,3-difluoro-4-hydroxypiperidine-1-carboxylate
tert-Butyl 3,3-difluoro-4-hydroxypiperidine-1-carboxylate
Formula: C₁₀H₁₇F₂NO₃
MW: 237.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16250751
CAS Number: 1209780-71-1