1,1'-(Piperidine-1,4-diyl)bis(2,2,2-trifluoroethanone) 95%
Supplier: Apollo Scientific
Synonyms: 1,1'-(Piperidine-1,4-diyl)bis(2,2,2-trifluoroethan-1-one)
PC430325-1G PC430325-250MG
APOSPC430325-1GEA 691 EUR
APOSPC430325-1G APOSPC430325-250MG
1,1'-(Piperidine-1,4-diyl)bis(2,2,2-trifluoroethanone) 95%
1,1'-(Piperidine-1,4-diyl)bis(2,2,2-trifluoroethanone)
Formula: C₉H₉F₆NO₂
MW: 277.164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11840379
CAS Number: 1159982-56-5