2,2,2-Trifluoro-4'-(trifluoromethyl)acetophenone 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2,2,2-Trifluoro-1-(4-(trifluoromethyl)phenyl)ethanone, 4-(Trifluoroacetyl)benzotrifluoride, 2,2,2-Trifluoro-1-[4-(trifluoromethyl)phenyl]ethan-1-one
PC4353-1G PC4353-25G PC4353-5G
APOSPC4353-1GEA 33.5 EUR
APOSPC4353-1G APOSPC4353-25G APOSPC4353-5G
2,2,2-Trifluoro-4'-(trifluoromethyl)acetophenone 97%
2,2,2-Trifluoro-4'-(trifluoromethyl)acetophenone
Formula: C₉H₄F₆O
Boiling Pt: 63…65 °C (15 mm Hg)
Density: 1.433 g/cm³ (25 °C)
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00061276
CAS Number: 74853-66-0
UN: 1993
ADR: 3,III