2'-Fluoro-4'-hydroxy-5'-methoxyacetophenone
Supplier: Apollo Scientific
Warning
Synonyms: 4-Acetyl-5-fluoro-2-methoxyphenol, 5-Acetyl-4-fluoro-2-hydroxyanisole, 1-(2-Fluoro-4-hydroxy-5-methoxyphenyl)ethanone, 1-(2-Fluoro-4-hydroxy-5-methoxyphenyl)ethan-1-one
PC4446-1G PC4446-5G PC4446-25G
APOSPC4446-1GEA 21.9 EUR
APOSPC4446-1G APOSPC4446-5G APOSPC4446-25G
2'-Fluoro-4'-hydroxy-5'-methoxyacetophenone
2'-Fluoro-4'-hydroxy-5'-methoxyacetophenone
Formula: C₉H₉FO₃
MW: 184.164 g/mol
Melting Pt: 113…114 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11101445
CAS Number: 1065076-49-4