α-Methyl-3-(trifluoromethyl)benzylamine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(3-(Trifluoromethyl)phenyl)ethanamine, 1-[3-(Trifluoromethyl)phenyl]ethylamine, 3-(1-Aminoethyl)benzotrifluoride
PC4459-1G PC4459-250MG
APOSPC4459-1GEA 49.3 EUR
APOSPC4459-1G APOSPC4459-250MG
α-Methyl-3-(trifluoromethyl)benzylamine 98%
α-Methyl-3-(trifluoromethyl)benzylamine
Formula: C₉H₁₀F₃N
MW: 189.178 g/mol
Boiling Pt: 69.4…70.8 °C (45 mm Hg)
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD04038877
CAS Number: 59382-36-4