N-[6-(Morpholin-4-yl)-4-(trifluoromethyl)pyridin-2-yl]ethane-1,2-diamine
Supplier: Apollo Scientific
PC446015-1G PC446015-5G
APOSPC446015-1GEA 244 EUR
APOSPC446015-1G APOSPC446015-5G
N-[6-(Morpholin-4-yl)-4-(trifluoromethyl)pyridin-2-yl]ethane-1,2-diamine
N-[6-(Morpholin-4-yl)-4-(trifluoromethyl)pyridin-2-yl]ethane-1,2-diamine
Formula: C₁₂H₁₇F₃N₄O
MW: 290.285 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11052353
CAS Number: 1053659-51-0