N-[6-(Pyrrolidin-1-yl)-4-(trifluoromethyl)pyridin-2-yl]ethane-1,2-diamine
Supplier: Apollo Scientific
PC446028-1G PC446028-5G
APOSPC446028-1GEA 244 EUR
APOSPC446028-1G APOSPC446028-5G
N-[6-(Pyrrolidin-1-yl)-4-(trifluoromethyl)pyridin-2-yl]ethane-1,2-diamine
N-[6-(Pyrrolidin-1-yl)-4-(trifluoromethyl)pyridin-2-yl]ethane-1,2-diamine
Formula: C₁₂H₁₇F₃N₄
MW: 274.285 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11052400
CAS Number: 1053657-86-5