5-Chloro-4-trifluoromethyl-pyridin-2-ol
Supplier: Apollo Scientific
PC446035-1G PC446035-5G
APOSPC446035-1GEA 308 EUR
APOSPC446035-1G APOSPC446035-5G
5-Chloro-4-trifluoromethyl-pyridin-2-ol
5-Chloro-4-trifluoromethyl-pyridin-2-ol
Formula: C₆H₃ClF₃NO
MW: 197.542 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11227192
CAS Number: 109919-31-5