2-Chloro-4,6-difluorophenol
Supplier: Apollo Scientific
Warning
PC4464-1G PC4464-250MG PC4464-5G
APOSPC4464-1GEA 58 EUR
APOSPC4464-1G APOSPC4464-250MG APOSPC4464-5G
2-Chloro-4,6-difluorophenol
2-Chloro-4,6-difluorophenol
Formula: C₆H₃ClF₂O
MW: 164.537 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11501032
CAS Number: 2267-99-4