6-Bromo-7-fluoroquinoline
Supplier: Apollo Scientific
|
Warning
|
PC448001-1G
PC448001-10G
PC448001-5G
APOSPC448001-1GEA
120
EUR
APOSPC448001-1G
APOSPC448001-10G
APOSPC448001-5G
6-Bromo-7-fluoroquinoline
6-Bromo-7-fluoroquinoline
Formula:
C₉H₅BrFN MW: 226.045 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13181511 CAS Number: 127827-52-5 |