Heptafluoro-2-naphthol 95%
Supplier: Apollo Scientific
Warning
PC4492-250MG PC4492-1G
APOSPC4492-250MGEA 243 EUR
APOSPC4492-250MG APOSPC4492-1G
Heptafluoro-2-naphthol 95%
Heptafluoro-2-naphthol
Formula: C₁₀HF₇O
MW: 270.103 g/mol
Melting Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00143689
CAS Number: 727-49-1