[3-(Perfluoroisopropoxy)prop-1-yl]trichlorosilane
Supplier: Apollo Scientific
Danger
Synonyms: {3-[(1,1,1,2,3,3,3-Heptafluoroprop-2-yl)oxy]prop-1-yl}trichlorosilane, Perfluoroisopropyl 3-(trichlorosilyl)propyl ether
PC4497-25G PC4497-5G
APOSPC4497-25GEA 0 EUR
APOSPC4497-25G APOSPC4497-5G
[3-(Perfluoroisopropoxy)prop-1-yl]trichlorosilane
[3-(Perfluoroisopropoxy)prop-1-yl]trichlorosilane
Formula: C₆H₆Cl₃F₇OSi
MW: 361.545 g/mol
Boiling Pt: 85…87 °C (35 mm Hg)
Density: 1.497 g/cm³ (25 °C)
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00045135
CAS Number: 15538-93-9
UN: 2987
ADR: 8,II