2,2-Difluoro-2-(2-methylphenyl)ethan-1-amine
Supplier: Apollo Scientific
PC450019-100MG PC450019-250MG
APOSPC450019-100MGEA 227 EUR
APOSPC450019-100MG APOSPC450019-250MG
2,2-Difluoro-2-(2-methylphenyl)ethan-1-amine
2,2-Difluoro-2-(2-methylphenyl)ethan-1-amine
Formula: C₉H₁₁F₂N
MW: 171.187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22124011
CAS Number: 1365807-92-6