3,4-Dichloro-1,1,1,3-tetrafluorobutane
Supplier: Apollo Scientific
PC450022-25G PC450022-5G
APOSPC450022-25GEA 372 EUR
APOSPC450022-25G APOSPC450022-5G
3,4-Dichloro-1,1,1,3-tetrafluorobutane
3,4-Dichloro-1,1,1,3-tetrafluorobutane
Formula: C₄H₄Cl₂F₄
MW: 198.974 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18074324
CAS Number: 1227947-61-6